Barium nitrate ≥99.95% (Ba basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-5624-50G 93-5624-10G
100208-194EA 142.95 USD
100208-194 100199-140
Barium nitrate ≥99.95% (Ba basis)
Barium nitrate
Formula: Ba(NO₃)₂
Melting Pt: 592 °C
Density: 3.230
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1,II
Merck Index: 13,00986

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR