Manganese(II) carbonate ≥90%

Supplier: STREM CHEMICALS INC MS

93-2533-100G 93-2533-500G
100199-026EA 46.71 USD
100199-026 100210-284
Manganese(II) carbonate ≥90%
Manganese (II) carbonate
Formula: CMnO₃
Melting Pt: 350 °C (dec.)
Density: 3.125
MDL Number: MFCD00011116
CAS Number: 598-62-9
EINECS: 209-942-9
Merck Index: 13,05749

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR