Copper(II)-2-ethylhexanoate (16 - 19% Cu) ≥98% solvent-free

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-Ethylhexanoic acid copper (II) salt, Copper(II) 2-ethylhexanoate, 2-Ethylcaproic acid copper (II) salt, Copper bis(2-ethylhexanoate)

29-2941-25G 29-2941-5G 29-2941-100G
100201-584EA 120.31 USD
100201-584 100198-738 100199-852
Copper(II)-2-ethylhexanoate (16 - 19% Cu) ≥98% solvent-free
Copper (II) 2-ethylhexanoate
Formula: C₁₆H₃₀CuO₄
Melting Pt: 252 °C
MDL Number: MFCD00015695
CAS Number: 149-11-1
EINECS: 205-731-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR