Tris(2-cyanoethyl)phosphine ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris (2-cyanoethyl)phosphine, 3,3',3''-Phosphinetriyltris(propiononitrile), 3,3',3''-Phosphinidynetris(propiononitrile)

15-7680-25G 15-7680-5G
100198-678EA 652.49 USD
100198-678 100198-680
Tris(2-cyanoethyl)phosphine ≥99%
Tris(2-cyanoethyl)phosphine

Phosphine

Formula: C₉H₁₂N₃P
Boiling Pt: 235 °C (0.9 mmHg)
Melting Pt: 97…98 °C
MDL Number: MFCD00013828
CAS Number: 4023-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR