Phosphonitrilic chloride trimer ≥98.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cyclic Phosphonitrilic Chloride Trimer, Hexachlorocyclotriphosphazene
15-4530-50G
15-4530-10G
100198-658EA
250.52
USD
100198-658
100209-730
Phosphonitrilic chloride trimer ≥98.5%
Phosphonitrilic chloride trimer
Phosphine
Formula:
Cl₆N₃P₃ Boiling Pt: 127 °C (12 mmHg) Melting Pt: 128.8 °C Density: 1,98 |
MDL Number:
MFCD00006474 CAS Number: 940-71-6 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...