Bis(p-sulfonatophenyl)phenylphosphine dihydrate dipotassium salt ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

15-0463-10G 15-0463-500MG 15-0463-2G
100198-618EA 2199.51 USD
100198-618 100209-672 100198-620
Bis(p-sulfonatophenyl)phenylphosphine dihydrate dipotassium salt ≥97%
Bis(p-sulfonatophenyl)phenylphosphine dihydrate dipotassium salt

Phosphine, Phosphine, Water Soluble

MDL Number: MFCD00269831
CAS Number: 151888-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR