(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, (S)-(-)-TolBINAP

15-0153-1G 15-0153-250MG
100206-312EA 101.91 USD
100206-312 100198-606
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl ≥98%
(S)-(-)-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl

BINAP, Phosphine

Formula: C₄₈H₄₀P₂
Melting Pt: 255…257 °C
MDL Number: MFCD01311709
CAS Number: 100165-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR