1,4,8,11-Tetraazacyclotetradecane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: CYCLAM

07-1959-1G 07-1959-250MG 07-1959-5G
100206-242EA 104.74 USD
100206-242 100198-504 100209-548
1,4,8,11-Tetraazacyclotetradecane ≥98%
1,4,8,11-Tetraazacyclotetradecane
Formula: C₁₀H₂₄N₄
Melting Pt: 185…186 °C
MDL Number: MFCD00005105
CAS Number: 295-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR