2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4,4'-Dicarboxy-2,2'-bipyridine, [2,2'-Bipyridine]-4,4'-dicarboxylic acid

07-0370-250MG 07-0370-5G 07-0370-1G
100198-498EA 94.83 USD
100198-498 100206-236 100206-234
2,2'-Bipyridine-4,4'-dicarboxylic acid ≥98%
2,2'-Bipyridine-4,4'-dicarboxylic acid
Formula: C₁₂H₈N₂O₄
MDL Number: MFCD00015430
CAS Number: 6813-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR