cis-Dichlorobis(triethylphosphine)platinum(II) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-7820-500MG 93-7820-2G
100198-460EA 236.37 USD
100198-460 100204-426
cis-Dichlorobis(triethylphosphine)platinum(II) ≥99%
cis-Dichlorobis(triethylphosphine)platinum(II)
Formula: PtCl₂[P(C₂H₅)₃]₂
MW: 502.304 g/mol
MDL Number: MFCD00058821
CAS Number: 15692-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR