Iron(II) sulfide ≥99% (Fe basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Ferrous sulphide, Ferrous sulfide
93-2640-100G
93-2640-500G
93-2640-2000G
100196-574EA
175.51
USD
100196-574
100198-346
100203-428
Iron(II) sulfide ≥99% (Fe basis)
Iron (II) sulphide
Formula:
FeS Melting Pt: 1195 °C Density: 4.694 g/cm³ (20 °C) |
MDL Number:
MFCD00011013 CAS Number: 1317-37-9 EINECS: 215-268-6 UN: 3077 ADR: 9,III Merck Index: 12,04106 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...