Potassium bromide ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

93-1909-500G 93-1909-100G
100198-320EA 121.72 USD
100198-320 100206-948
Potassium bromide ≥99% ACS
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C
Melting Pt: 730 °C
Density: 2.75
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR