Iron(III) meso-tetraphenylporphine chloride

Supplier: STREM CHEMICALS INC MS

Synonyms: Iron(III) Tetraphenylporphyrin chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride, meso-Tetraphenylporphyrin iron(III) chloride, iron(III) chloride TPP, 21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride

26-2880-500MG 26-2880-2G
100198-160EA 165.6 USD
100198-160 100200-910
Iron(III) meso-tetraphenylporphine chloride
Iron(III) meso-tetraphenylporphine chloride

Porphine

Formula: C₄₄H₂₈ClFeN₄
MDL Number: MFCD00012152
CAS Number: 16456-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR