Trimethylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Methyl phosphine

Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC)

15-6502-25G
100198-140EA 702.03 USD
100198-140
Trimethylphosphine ≥98%
Trimethylphosphine

Phosphine

Formula: C₃H₉P
Boiling Pt: 38…40 °C
Melting Pt: –85 °C
Density: 0,748
Flash Pt: –22 °F
MDL Number: MFCD00008510
CAS Number: 594-09-2
EINECS: 209-823-1
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR