1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

09-4750-25G 09-4750-5G
100198-098EA 186.83 USD
100198-098 100201-404
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
1,1,1,3,3,3-Hexafluoro-2-propanol

Organo Fluorine

Formula: C₃H₂F₆O
Boiling Pt: 58 °C
Melting Pt: –4 °C
Density: 1.596
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,II
Merck Index: 13,09598

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR