Tris(dibenzylideneacetone)dipalladium(0)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]
46-3000-5G
46-3000-1G
100198-026EA
389.23
USD
100198-026
100201-686
Tris(dibenzylideneacetone)dipalladium(0)
Tris(dibenzylideneacetone)dipalladium(0)
Formula:
C₅₁H₄₂O₃Pd₂ Melting Pt: 152…155 °C |
MDL Number:
MFCD00013310 CAS Number: 51364-51-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...