Vanadium ≥99.5%, foil

Supplier: STREM CHEMICALS INC MS

23-0050-100X50MM 23-0050-100X200MM
100198-020EA 246.28 USD
100198-020 100198-018
Vanadium ≥99.5%, foil
Vanadium
Formula: V
Boiling Pt: 3000 °C
Melting Pt: 1717 °C
Density: 6.11
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR