Antimony(III) chloride ≥99.999% (Sb basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Antimony trichloride

97-0373-25G
100197-980EA 604.37 USD
100197-980
Antimony(III) chloride ≥99.999% (Sb basis), PURATREM, Electronic Grade
Antimony (III) chloride

Halides for CVD & ALD

Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 283 °C
Melting Pt: 73.4 °C
Density: 3.140
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 13,00713

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR