Samarium ≥99.9%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-6235-10G 93-6235-50G
100197-924EA 186.83 USD
100197-924 100197-926
Samarium ≥99.9%, powder
Samarium
Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900
Melting Pt: 1076.85
Density: 7.4
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
UN: 3089
ADR: 4 III
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR