Zinc hexafluoro-2,4-pentanedionate dihydrate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc hexafluoroacetylacetonate hydrate

93-3035-1G 93-3035-5G
100197-816EA 86.34 USD
100197-816 100210-348
Zinc hexafluoro-2,4-pentanedionate dihydrate ≥98%
Zinc hexafluoro-2,4-pentanedionate dihydrate
Formula: C₁₀H₂F₁₂O₄Zn.2H₂O
Melting Pt: 157…158 °C
MDL Number: MFCD00150577
CAS Number: 16743-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR