Zinc fluoride, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-3011-100G 93-3011-25G
100202-548EA 209.48 USD
100202-548 100197-814
Zinc fluoride, anhydrous ≥99%
Zinc fluoride
Formula: F₂Zn
Boiling Pt: ~1500 °C
Melting Pt: 872 °C
Density: 4.95
MDL Number: MFCD00011298
CAS Number: 7783-49-5
EINECS: 232-001-9
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR