Aluminium perchlorate nonahydrate, Reagent Grade

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminum perchlorate nonahydrate

93-1340-250G 93-1340-50G
100197-694EA 131.63 USD
100197-694 100198-976
Aluminium perchlorate nonahydrate, Reagent Grade
Aluminium perchlorate nonahydrate
Formula: AlCl₃O₁₂·9H₂O
Density: 2.0
MDL Number: MFCD00149130
CAS Number: 81029-06-3
UN: 1481
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR