Sodium stearate, Technical Grade

Supplier: STREM CHEMICALS INC MS

Synonyms: Stearate sodium, Stearic acid sodium salt

93-1180-1000G
100197-672EA 66.52 USD
100197-672
Sodium stearate, Technical Grade
Sodium stearate
Formula: C₁₈H₃₅NaO₂
Melting Pt: 288 °C
Density: 1.103 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036404
CAS Number: 822-16-2
EINECS: 212-490-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR