Hafnium(IV) tert-butoxide ≥99.9% (Hf basis) <1.5%-Zr

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hafnium(IV) t-butoxide, Hafnium(IV)-tert-butanolate

72-5800-2G 72-5800-10G
100201-722EA 144.37 USD
100201-722 100197-586
Hafnium(IV) tert-butoxide ≥99.9% (Hf basis) <1.5%-Zr
Hafnium(IV) tert-butoxide

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₁₆H₃₆HfO₄
Boiling Pt: 90 °C (5 mmHg)
Melting Pt: 8 °C
Density: 1.166
MDL Number: MFCD00070458
CAS Number: 2172-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR