Lanthanum(III) nitrate hexahydrate ≥99.999% (La basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

57-5732-100G 57-5732-25G
100197-568EA 257.6 USD
100197-568 100206-772
Lanthanum(III) nitrate hexahydrate ≥99.999% (La basis), PURATREM
Lanthanum (III) nitrate hexahydrate
Formula: La(NO₃)₃·6H₂O
Boiling Pt: 126 °C (dec.)
Melting Pt: 40 °C
Density: 2.347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,II
Merck Index: 14,05363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR