Tin(IV) oxide ≥99.998% (Sn basis), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

50-2500-5G 50-2500-25G
100203-226EA 164.18 USD
100203-226 100197-556
Tin(IV) oxide ≥99.998% (Sn basis), PURATREM
Tin (IV) oxide
Formula: O₂Sn
Boiling Pt: ∼2500 °C (1013 hPa)
Melting Pt: 1630 °C
Density: 7.0
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR