Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

46-2150-5G 46-2150-1G
100197-540EA 227.88 USD
100197-540 100201-684
Tetrakis(triphenylphosphine)palladium(0) ≥99% (99.9+%-Pd)
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
Melting Pt: 103…107 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR