Selenium ≥99.99%, powder

Supplier: STREM CHEMICALS INC MS
Danger

34-0090-500G 34-0090-100G
100197-498EA 968.12 USD
100197-498 100197-496
Selenium ≥99.99%, powder
Selenium
Formula: Se
Boiling Pt: 690 °C
Melting Pt: 170…217 °C
Density: 4.81
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3283
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR