Zinc sulfide ≥99.99% (Zn basis), fused granules, PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

30-3024-5G 30-3024-25G 30-3024-100G
100197-490EA 35.38 USD
100197-490 100198-176 100198-744
Zinc sulfide ≥99.99% (Zn basis), fused granules, PURATREM
Zinc sulphide
Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.10
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR