Tricarbonyl(cyclooctatetraene)iron ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cyclooctatetraene iron tricarbonyl, Cyclooctatetraene Iron Tricarbonyl

26-0875-1G 26-0875-5G
100197-464EA 291.57 USD
100197-464 100197-466
Tricarbonyl(cyclooctatetraene)iron ≥98%
Tricarbonyl(cyclooctatetraene)iron

Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₁H₈FeO₃
Melting Pt: 93…95 °C
MDL Number: MFCD00001755
CAS Number: 12093-05-9
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR