Potassium tris(1-pyrazolyl)borohydride hydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

19-3600-5G 19-3600-1G
100197-436EA 527.94 USD
100197-436 100197-434
Potassium tris(1-pyrazolyl)borohydride hydrate ≥98%
Potassium tris(1-pyrazolyl)borohydride hydrate
Formula: C₉H₁₀BKN₆·xH₂O
Melting Pt: 185…189 °C
MDL Number: MFCD03414149
CAS Number: 18583-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR