Cyclohexyldiphenylphosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Diphenylcyclohexylphosphine
15-0900-5G
15-0900-1G
100207-344EA
246.28
USD
100207-344
100197-398
Cyclohexyldiphenylphosphine ≥98%
Cyclohexyldiphenylphosphine
Phosphine
Formula:
C₁₈H₂₁P MW: 268.33 g/mol Melting Pt: 58…62 °C |
MDL Number:
MFCD00046360 CAS Number: 6372-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...