Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Al(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)aluminum
13-5000-1G
13-5000-5G
13-5000-25G
100197-370EA
63.69
USD
100197-370
100197-374
100197-372
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇AlO₆ Boiling Pt: >400 °C/ 150 °C (0.01 mmHg) (dec./ subl.) Melting Pt: 255…258 °C |
MDL Number:
MFCD00058918 CAS Number: 14319-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...