Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)

Supplier: STREM CHEMICALS INC MS

Synonyms: Al(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)aluminum

13-5000-1G 13-5000-5G 13-5000-25G
100197-370EA 63.69 USD
100197-370 100197-374 100197-372
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium ≥99% (99.9%-Al)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇AlO₆
Boiling Pt: >400 °C/ 150 °C (0.01 mmHg) (dec./ subl.)
Melting Pt: 255…258 °C
MDL Number: MFCD00058918
CAS Number: 14319-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR