1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol

09-6147-5G 09-6147-25G
100197-354EA 70.77 USD
100197-354 100209-624
1H,1H,2H,2H-Tridecafluoro-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluoro-1-octanol

Organo Fluorine

Formula: C₈H₅F₁₃O
MW: 364.11 g/mol
Boiling Pt: 91…93 °C (36 mmHg)
Density: 1.651
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR