EDTA (ethylenediamine tetraacetic acid) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-0573-500G 07-0573-2000G
100197-324EA 140.12 USD
100197-324 100210-962
EDTA (ethylenediamine tetraacetic acid) ≥99%
EDTA (Ethylenediamine tetraacetic acid)
Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 245 °C (dec.)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR