Carbon nanotube multi-walled, as produced cathode deposit

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: CNT

06-0504-5G 06-0504-1G
100197-304EA 274.58 USD
100197-304 100210-954
Carbon nanotube multi-walled, as produced cathode deposit
Carbon nanotube

Carbon Nanotubes, Nanomaterials

Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR