Charcoal activated

Supplier: STREM CHEMICALS INC MS
Warning

06-0100-1000G 06-0100-300G
100210-946EA 144.37 USD
100210-946 100197-300
Charcoal activated
Charcoal activated
Formula: C
Boiling Pt: 4200 °C
Melting Pt: 3652…3697 °C (subl.)
Density: 1.8…2.1 (amorphous)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR