Hafnium ≥99.6% (Zr-2.2%), powder

Supplier: STREM CHEMICALS INC MS
Danger

93-7227-10G 93-7227-2G 93-7227-50G
100197-268EA 154.28 USD
100197-268 100205-750 100207-148
Hafnium ≥99.6% (Zr-2.2%), powder
Hafnium
Formula: Hf
Boiling Pt: 4602 °C
Melting Pt: 2227 °C
Density: 13.31
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4
UN: 2545
ADR: 4.2,II
Merck Index: 12,04618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR