Dysprosium trifluoride, anhydrous ≥99.9% (dye basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6609-100G 93-6609-25G
100208-590EA 1416.8 USD
100208-590 100197-262
Dysprosium trifluoride, anhydrous ≥99.9% (dye basis)
Dysprosium trifluoride
Formula: DyF₃
Boiling Pt: > 2200 °C
Melting Pt: 1360 °C
Density: 5.948 g/cm³ (1149 °C)
MDL Number: MFCD00016071
CAS Number: 13569-80-7
EINECS: 236-992-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR