Aluminium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

93-1379-100MM 93-1379-500MM
100197-196EA 157.11 USD
100197-196 100196-496
Aluminium ≥99.9%, foil
Aluminium
Formula: Al
Boiling Pt: 2056 °C
Melting Pt: 660 °C
Density: 2.702
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR