Tungsten(IV) chloride ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tungsten tetrachloride

74-2348-5G 74-2348-25G
100197-164EA 179.75 USD
100197-164 100200-298
Tungsten(IV) chloride ≥97%
Tungsten (IV) chloride
Formula: Cl₄W
Density: 4.624
MDL Number: MFCD00012027
CAS Number: 13470-13-8
UN: 1759
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR