Aluminium trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Aluminum tris(trifluoromethanesulfonate), Aluminum trifluoromethanesulphonate, Aluminum trifluoromethanesulfonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)

13-1500-10G 13-1500-50G
100200-846EA 162.77 USD
100200-846 100197-062
Aluminium trifluoromethanesulfonate ≥99%
Aluminium trifluoromethanesulphonate

Metal Triflate

Formula: C₃AlF₉O₉S₃
Melting Pt: Dec.
MDL Number: MFCD00143596
CAS Number: 74974-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR