Hexachloroplatinic(IV) acid hexahydrate ≥99.9% (Pt basis) (38-40% Pt)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate
78-0200-1G
78-0200-5G
78-0200-25G
100204-106EA
124.55
USD
100204-106
100197-030
100200-490
Hexachloroplatinic(IV) acid hexahydrate ≥99.9% (Pt basis) (38-40% Pt)
Hexachloroplatinic (IV) acid hexahydrate
Formula:
H₂PtCl₆·6H₂O Melting Pt: 60 °C Density: 2.434 g/cm³ (20 °C) |
MDL Number:
MFCD00149910 CAS Number: 18497-13-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III Merck Index: 13,07609 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...