Bismuth ≥99.9999%, rods

Supplier: STREM CHEMICALS INC MS

83-0080-25G 83-0080-100G
100196-976EA 510.95 USD
100196-976 100210-916
Bismuth ≥99.9999%, rods
Bismuth
Formula: Bi
Boiling Pt: 1560 °C
Melting Pt: 271.3 °C
Density: 9.80
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR