Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: [Ba(TMHD)2], Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium hydrate

56-8500-5G 56-8500-25G 56-8500-1G
100207-416EA 222.22 USD
100207-416 100196-958 100197-566
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₂₂H₃₈BaO₄·H₂O
Boiling Pt: 285 °C/ 225 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 195…200 °C
MDL Number: MFCD06200255
CAS Number: 17594-47-7
UN: 1564
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR