Bismuth(III) oxide ≥99.999% (Bi basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
93-8319-500G
93-8319-100G
93-8319-25G
100199-204EA
673.72
USD
100199-204
100205-800
100196-842
Bismuth(III) oxide ≥99.999% (Bi basis), PURATREM
Bismuth (III) oxide
Formula:
BiO₃ Boiling Pt: 1890 °C Melting Pt: 817 °C Density: 8.9 |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 13,01273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...