Samarium ≥99.9%, chips

Supplier: STREM CHEMICALS INC MS

93-6236-50G 93-6236-10G
100196-798EA 513.78 USD
100196-798 100196-796
Samarium ≥99.9%, chips
Samarium
Formula: Sm
MW: 150.36 g/mol
Boiling Pt: 1900
Melting Pt: 1076.85
Density: 7.4
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR