Samarium(III) oxalate hydrate ≥99.9% (Sm basis)
Supplier: STREM CHEMICALS INC MS
93-6209-10G
93-6209-50G
100196-792EA
77.85
USD
100196-792
100198-428
Samarium(III) oxalate hydrate ≥99.9% (Sm basis)
Samarium(III) oxalate hydrate
Formula:
C₆O₁₂Sm₂.xH₂O |
MDL Number:
MFCD00150491 CAS Number: 14175-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...