Cesium acetate ≥99.9% (Cs basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Acetic acid cesium salt

93-5510-10G 93-5510-50G
100196-754EA 77.85 USD
100196-754 100196-756
Cesium acetate ≥99.9% (Cs basis)
Cesium acetate
Formula: H₃CCOOCs
Melting Pt: 194 °C
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR